Order Entry
United States
Orders LinkContactUsLinkComponent
D-(-)-Glutamic acid 99+%

D-(-)-Glutamic acid 99+%

Glass bottle for solids, 5 g
Catalog # TS22789-0050
Supplier #:  22789-0050
D-(-)-Glutamic acid 99+%
Catalog # TS22789-0050
CAS Number:  6893-26-1
D-(-)-Glutamic acid 99+%
D-(-)-Glutamic acid 99+%

Glass bottle for solids, 5 g

CAS Number:  6893-26-1
Price
$49.70
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  • CAS Number:6893-26-1
  • Formula:C₅H₉NO₄
  • MDL Number:MFCD00063112
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:200…202 °C
  • EINECS:230-000-8
Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.2 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≤1 % L-content
  • Specific optical rotation:
    -30.5° to -32.2° (20°C, 589 nm) (c=10, 2 N HCl)
  • Titration with NaOH:
    99.0 to 101.0 %
  • Other amino acids:
    ≤0.5 % (TLC)

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.