D-(+)-Mannose 99+%
Plastic drum, 2.5 kg
Catalog # TS15060-0025
Supplier: Thermo Scientific Chemicals
: 15060-0025
D-(+)-Mannose 99+%
Catalog # TS15060-0025
Supplier: Thermo Scientific Chemicals
CAS Number: 3458-28-4
Jump to:
Product Details & Documents
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00064122
- Molecular Weight:180.16 g/mol
- Merck Index:13,05772
- Melting Pt:133 °C
- Storage Temperature:Ambient
- EINECS:222-392-4
Specifications
Specification Test Results
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.45 %
- HPLC:≥99.0 %
- Appearance of solution:(10 % in water) Clear
- Bacterial endotoxins:(LAL-test) 0.5 E.U. in 1 ml of a 0.1% soln.
- Specific optical rotation:+13.5° to +14.9° (20°C, 589 nm) (c=10, water) (after 24 hours, on dry substance)
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Plastic drum
- Size:2.5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



