Orders LinkContactUsLinkComponent
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered, Cube, 5 gallon
Catalog # 76548-090
Supplier:   StatLab
Supplier #:  BFZ-5
Zinc Formalin Buffered , Statlab
Catalog # 76548-090
Supplier:  StatLab
CAS Number:  
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered, Cube, 5 gallon
CAS Number:  
Price
$93.96
Log in for availability
Each (5US Gal)
Product Details & Documents
Ready-to-use Zinc Formalin solution.
  • Especially recommended for use in immunohistochemistry
  • Zinc formalin preserves more immunoreactivity than standard NBF
Part of the StatLab line of StatClick 10% NBF Prefills.
Manufactured in the USA
Specifications
Specifications
  • Pack type:
    Cube
  • Description:
    Zinc Formalin Buffered
  • Size:
    5 gallon