Orders LinkContactUsLinkComponent
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered, 1 gallon
Catalog # 76548-088
Supplier #:  BFZ-1
Zinc Formalin Buffered , Statlab
Catalog # 76548-088
Supplier:  
CAS Number:  
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered, 1 gallon
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FORMALIN ZINC BUFFERED 1GAL Catalog #76548-089
Product 76548-089 is a replacement for 76548-088 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Ready-to-use Zinc Formalin solution.
  • Especially recommended for use in immunohistochemistry
  • Zinc formalin preserves more immunoreactivity than standard NBF
Part of the StatLab line of StatClick 10% NBF Prefills.
Manufactured in the USA
Specifications
Specifications
  • Description:
    Zinc Formalin Buffered
  • Size:
    1 gallon