Order Entry
United States
Orders LinkContactUsLinkComponent
Zinc Formalin Buffered , Statlab

Zinc Formalin Buffered , Statlab

Zinc Formalin Buffered, 60 ml
Catalog # 76548-084
Supplier:   StatLab
Supplier #:  BFZ0230
Zinc Formalin Buffered , Statlab
Catalog # 76548-084
Supplier:  StatLab
CAS Number:  
Zinc Formalin Buffered , Statlab
Zinc Formalin Buffered , Statlab

Zinc Formalin Buffered, 60 ml

CAS Number:  
Price
$93.32
Log in for availability
Case of 96 (60ml)
Product Details & Documents

Ready-to-use Zinc Formalin solution.

  • Especially recommended for use in immunohistochemistry
  • Zinc formalin preserves more immunoreactivity than standard NBF

Part of the StatLab line of StatClick 10% NBF Prefills.

Manufactured in the USA

Specifications

Specifications

  • Description:
    Zinc Formalin Buffered
  • Size:
    60 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.