Orders LinkContactUsLinkComponent
Immunocal™ Decalcifier, StatLab
Immunocal™ Decalcifier, StatLab
Immunocal™ Decalcifier, 1 Gallon
Catalog # 76548-020
Supplier:   StatLab
Supplier #:  1414-1
Immunocal™ Decalcifier, StatLab
Catalog # 76548-020
Supplier:  StatLab
CAS Number:  
Immunocal™ Decalcifier, StatLab
Immunocal™ Decalcifier, StatLab
Immunocal™ Decalcifier, 1 Gallon
CAS Number:  
Price
$75.38
Log in for availability
Each (1US Gal)
Product Details & Documents
Very mild formic acid decalcifier formulated from semiconductor grade formic acid and highly purified reagent grade water. Ideal for decalcifying bone for immunohistochemistry. Will not destroy antigen sites during decalcification and can safely be used to demonstrate antigens in decalcified bone.
Packaging: Manufactured in the USA
Documents
Specifications
Specifications
  • Description:
    Immunocal™ Decalcifier
  • Size:
    1 Gallon