Order Entry
United States
Orders LinkContactUsLinkComponent
Formical-4™ Decalcifier, StatLab

Formical-4™ Decalcifier, StatLab

Formical-4™ Decalcifier, 32 oz
Catalog # 76548-010
Supplier:   StatLab
Supplier #:  1214-32
Formical-4™ Decalcifier, StatLab
Catalog # 76548-010
Supplier:  StatLab
CAS Number:  
Formical-4™ Decalcifier, StatLab
Formical-4™ Decalcifier, StatLab

Formical-4™ Decalcifier, 32 oz

CAS Number:  
Price
$35.34
Log in for availability
Each (32oz)
Product Details & Documents

Fixative decalcifier can save time. Fresh tissue can be placed directly into Formical-4™ to fix and decalcify simultaneously. Formulated for use on large dense samples that normally take several days to decalcify. In most cases, poor decalcification is due to poor fixation as large, dense bone usually does not fix properly.

As Formical-4™ begins to decalcify large specimens, it allows better penetration of formaldehyde. Therefore, cortical bone, femoral heads, and teeth will not only decalcify better, they will fix better. Formical-4 has also proven to be a smart choice for IHC and ISH.

Packaging: Manufactured in the USA

Specifications

Specifications

  • Description:
    Formical-4™ Decalcifier
  • Size:
    32 oz

Specifications