Orders LinkContactUsLinkComponent
Formical-4™ Decalcifier, StatLab
Formical-4™ Decalcifier, StatLab
Formical-4™ Decalcifier, 1 Gallon
Catalog # 76548-008
Supplier #:  1214-1
Formical-4™ Decalcifier, StatLab
Catalog # 76548-008
Supplier:  
CAS Number:  
Formical-4™ Decalcifier, StatLab
Formical-4™ Decalcifier, StatLab
Formical-4™ Decalcifier, 1 Gallon
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
DECALCIFIER FORMICAL-4 1GAL Catalog #76548-009
Product 76548-009 is a replacement for 76548-008 but varies by Brand, Packaging or Quantity.
DECALCIFIER FORMICAL-4 5 GAL CUBE Catalog #76548-012
Product 76548-012 is a replacement for 76548-008 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Fixative decalcifier can save time. Fresh tissue can be placed directly into Formical-4™ to fix and decalcify simultaneously. Formulated for use on large dense samples that normally take several days to decalcify. In most cases, poor decalcification is due to poor fixation as large, dense bone usually does not fix properly.
As Formical-4™ begins to decalcify large specimens, it allows better penetration of formaldehyde. Therefore, cortical bone, femoral heads, and teeth will not only decalcify better, they will fix better. Formical-4 has also proven to be a smart choice for IHC and ISH.
Packaging: Manufactured in the USA
Specifications
Specifications
  • Description:
    Formical-4™ Decalcifier
  • Size:
    1 Gallon