Order Entry
United States
Orders LinkContactUsLinkComponent
Fixative Rapid Differential Stain Hema-Diff™, Statlab

Fixative Rapid Differential Stain Hema-Diff™, Statlab

#1 Fixative Rapid Differential Stain Hema-Diff™, 16 oz
Catalog # 76547-990
Supplier:   StatLab
Supplier #:  11985-16
Fixative Rapid Differential Stain Hema-Diff™, Statlab
Catalog # 76547-990
Supplier:  StatLab
CAS Number:  
Fixative Rapid Differential Stain Hema-Diff™, Statlab
Fixative Rapid Differential Stain Hema-Diff™, Statlab

#1 Fixative Rapid Differential Stain Hema-Diff™, 16 oz

CAS Number:  
Price
$50.45
Log in for availability
Each (16oz)
Product Details & Documents

The original StatLab best-selling rapid stain for the past 40 years. Three-step procedure for differential staining of blood smears, used extensively in FNA cytology.

  • Entire procedure takes 30 seconds as air dried specimens are fixed in Solution I, then dipped in Solutions II and III
  • Excellent for FNA cytology, differential staining of blood smears, microbiological organisms, H. pylori, and bone marrow specimens
  • Versatile and very economical
  • Components sold separately or as a kit
  • Can also be used as a special stain for Helicobacter pylori. 16 oz Hema-Diff #1 Fixative

Part of the StatLab Hema-Diff Rapid Differential Stain Line.

Manufactured in the USA

Specifications

Specifications

  • Description:
    #1 Fixative Rapid Differential Stain Hema-Diff™
  • Size:
    16 oz

Specifications