Fixative Rapid Differential Stain Hema-Diff™, Statlab
#1 Fixative Rapid Differential Stain Hema-Diff™, 1 gallon
Catalog # 76547-988
Supplier #: 11985-1
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
STAIN CYTO FIXATIVE HEMA-DIFF NO. 1 1GAL Catalog #76547-989
Product 76547-989 is a replacement for 76547-988 but varies by Brand, Packaging or Quantity.
Product Details & Documents
The original StatLab best-selling rapid stain for the past 40 years. Three-step procedure for differential staining of blood smears, used extensively in FNA cytology.
- Entire procedure takes 30 seconds as air dried specimens are fixed in Solution I, then dipped in Solutions II and III
- Excellent for FNA cytology, differential staining of blood smears, microbiological organisms, H. pylori, and bone marrow specimens
- Versatile and very economical
- Components sold separately or as a kit
- Can also be used as a special stain for Helicobacter pylori. 16 oz Hema-Diff #1 Fixative
Part of the StatLab Hema-Diff Rapid Differential Stain Line.
Manufactured in the USA
Specifications
Specifications
- Description:#1 Fixative Rapid Differential Stain Hema-Diff™
- Size:1 gallon