STABILIZER PROTEOMIC PROT1 100 TEST
Catalog # MSPP-501351689
Supplier: SMART TUBE INC MS
STABILIZER PROTEOMIC PROT1 100 TEST
Catalog # MSPP-501351689
Supplier: SMART TUBE INC MS
Supplier Number: 501351689
Frequently Bought Together
Disk Dispenser
Supplier: BBL
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International