ANTISERUM SHEEP RED 5ML 5X10-8/ML VOLUME
Catalog # MSPP-B200
Supplier: COMPTECH MS
ANTISERUM SHEEP RED 5ML 5X10-8/ML VOLUME
Catalog # MSPP-B200
Supplier: COMPTECH MS
Supplier Number: B200
Frequently Bought Together
NeoPro® Powder-Free Neoprene Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
VWR® Talon® Lab Frame Feet
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Derma Free® Vinyl Gloves, Microflex®
Supplier: Ansell Healthcare
neodisher® Detergents, Miele
Supplier: Miele
SYRINGE INSULIN 29GX1/2IN 3/10ML CS300
Supplier: Cardinal Health
VWR® Sleeve Stoppers
Supplier: VWR International
VWR® Premium Aluminum Foil
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BD Trypticase™ Soy Agar, BD Diagnostics
Supplier: BD
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Anti-C3 Goat Polyclonal Antibody
Supplier: MP Biomedicals
Neutalizer, LancerAcid Rinse, LCA-A, Lancer
Supplier: GETINGE USA SALES, LLC. MS
Lok-Tight™ Contact Plates for Surface Sampling, Hardy Diagnostics
Supplier: Hardy Diagnostics
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Sodium chloride 0.9% in water normal saline solution
Supplier: G-Biosciences
BUFFER GVB WITH CA/MG FOR ASSAYS 1 LITER
Supplier: COMPTECH MS