Vee Gee Snap Cap Replacements
#2, 25 ml flasks (76546-118)
Catalog # 76546-718
Supplier: Heathrow Scientific
Supplier #: SC#2
Jump to:
Product Details & Documents
Replacement snap-caps.
- For SIBATA volumetric flasks
Sold by the each.
Specifications
- Cap Size:#2
- Fits:25 ml flasks (76546-118)