Order Entry
United States
Orders LinkContactUsLinkComponent
Wide-Range Specific Gravity Hydrometers

Wide-Range Specific Gravity Hydrometers

1.800...2.050, 165 mm (6.5"), 0.005
Catalog # 76546-434
Supplier:   Heathrow Scientific
Supplier #:  6604-5
Wide-Range Specific Gravity Hydrometers
Catalog # 76546-434
Wide-Range Specific Gravity Hydrometers
Wide-Range Specific Gravity Hydrometers

1.800...2.050, 165 mm (6.5"), 0.005

Price
$29.06
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

These wide range specific gravity hydrometers are ideal to use when you aren't sure of the approximate value of your liquid. These refractometers can also be used for measuring multiple liquids with different specific gravity amounts.

All of our specific gravity hydrometers are calibrated at 60 °F.

Specifications
  • Length:
    165 mm (6.5")
  • Range:
    1.800...2.050
  • Subdivisions:
    0.005
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.