Order Entry
United States
Orders LinkContactUsLinkComponent
Hydrometer Jars

Hydrometer Jars

600 ml, Without graduation lines, max. hydrometer length 390 mm (15.4")
Catalog # 76546-102
Supplier:   Heathrow Scientific
Supplier #:  21050-600
Hydrometer Jars
Catalog # 76546-102
Hydrometer Jars
Hydrometer Jars

600 ml, Without graduation lines, max. hydrometer length 390 mm (15.4")

Price
$18.66
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Hydrometer jars are made of borosilicate glass and feature a beaded rim with a circular base.

  • Available in a variety of capacities

All hydrometer jars are sized properly to eliminate surface tension effects that lead to inaccurate readings. Models 76546-096 have a graduation line at 1000 ml for precise filling and is designed for use as a sedimentation cylinder.

Specifications
  • Capacity:
    600 ml
  • Description:
    Without graduation lines
  • Use with:
    max. hydrometer length 390 mm (15.4")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.