Order Entry
United States
Orders LinkContactUsLinkComponent
Hydrometer Jars

Hydrometer Jars

500 ml, Without graduation lines, max. hydrometer length 360 mm (14.2")
Catalog # 76546-100
Supplier:   Heathrow Scientific
Supplier #:  21050-500
Hydrometer Jars
Catalog # 76546-100
Hydrometer Jars
Hydrometer Jars

500 ml, Without graduation lines, max. hydrometer length 360 mm (14.2")

Price
$18.02
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Hydrometer jars are made of borosilicate glass and feature a beaded rim with a circular base.

  • Available in a variety of capacities

All hydrometer jars are sized properly to eliminate surface tension effects that lead to inaccurate readings. Models 76546-096 have a graduation line at 1000 ml for precise filling and is designed for use as a sedimentation cylinder.

Specifications
  • Capacity:
    500 ml
  • Description:
    Without graduation lines
  • Use with:
    max. hydrometer length 360 mm (14.2")