SimLabSolutions Injection Trainer
Injection trainer, large
Catalog # 470338-474
Supplier: DIAMEDICAL SE
Supplier #: MS153902
Jump to:
Product Details & Documents
Easily transportable injection training.
- Real feel for skin puncture resistance
- Self-sealing
- Foam core may be injected with water
- Drains easily and dries without deteriorating
- Removable top layer is suitable for practicing intradermal injections
The SimLabSolutions injection trainers are a convenient and easy way to practice different injection techniques. The smaller pad is a perfect size for an individual student, while the larger size has a higher surface area for longer classroom durability. The container protects the pad between uses, as well as acting as a shield from accidental over sticks. The pad accommodates both dry and wet practice. The foam core is easily drained and dried.
Documents
Injection Trainers Demo
Videos
Specifications
- Description:Injection trainer, large