Orders LinkContactUsLinkComponent
Discover Simulation Headwall Packages
Discover Simulation Headwall Packages
60" Discover Headwall, 60"
Catalog # 470338-424
Supplier:   DIAMEDICAL SE
Supplier #:  HW020805
Discover Simulation Headwall Packages
Catalog # 470338-424
Supplier:  DIAMEDICAL SE
Discover Simulation Headwall Packages
Discover Simulation Headwall Packages
60" Discover Headwall, 60"
Price
$7,071.49
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Simulation headwall with air.
  • Multiple sizes available
  • Wall mounted; mounting strip included
  • Nurse call facade with code blue button and hand pendant
  • Includes silent air compressor
  • Includes complete accessories package
The Functioning Wall Mounted Discover Headwall Package simulates oxygen, air, and suction with one source of air by connecting to an external air compressor. This solution is thousands of dollars less than a traditional centralized system with air, oxygen, and vacuum.
Please see product resources for contents.
Ordering information: Default color is off white. Other colors, sizes, and configurations are available. Please consult with your sales representative for more information.
Caution: This is a non-functional headwall intended for simulation training.
Specifications
  • Description:
    60" Discover Headwall
  • Length:
    60"