Orders LinkContactUsLinkComponent
Discover Simulation Headwall Packages
48" Discover Headwall with Light, 48"
  470338-358
 :  HW020804
Discover Simulation Headwall Packages
  470338-358
Discover Simulation Headwall Packages
Discover Simulation Headwall Packages
48" Discover Headwall with Light, 48"
$6,733.39
Each
Simulation headwall with air.
  • Multiple sizes available
  • Wall mounted; mounting strip included
  • Nurse call facade with code blue button and hand pendant
  • Includes silent air compressor
  • Includes complete accessories package
The Functioning Wall Mounted Discover Headwall Package simulates oxygen, air, and suction with one source of air by connecting to an external air compressor. This solution is thousands of dollars less than a traditional centralized system with air, oxygen, and vacuum.
Please see product resources for contents.
 : Default color is off white. Other colors, sizes, and configurations are available. Please consult with your sales representative for more information.
 : This is a non-functional headwall intended for simulation training.
  • Description:
    48" Discover Headwall with Light
  • Length:
    48"