PL Multisolvent, Agilent Technologies
PL Multisolvent 30, Analytical, PL Multisolvent 30, 2.7 µm, 4.60 mm, 150 mm, Fully Porous
Catalog # AGPL1515-3323
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: PL1515-3323
PL Multisolvent, Agilent Technologies
Catalog # AGPL1515-3323
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
PL Multisolvent GPC columns use polymer-coated silica to operate in a wide range of solvents.
- The PL Multisolvent has the efficiency and robustness of silica with a special coating to eliminate ‘sticking’
- Silica backbone provides the high strength necessary for high-efficiency particles and the rigid structure that eliminates pore swelling and collapse with solvent switching
- High-resolution separation of polymers up to 100,000 Da, as well as additives, oligomers, and plasticizers
The PL Multisolvent family uses a revolutionary process to coat high-pore volume silica in an inert polymer. Columns separate low and moderate molecular weight polymers in solvents ranging from water to tetrahydrofuran (THF) without secondary interactions. The silica backbone provides excellent resolution and eliminates degradation caused by swelling during solvent transfer.
Specifications
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:2.7 µm
- Description:PL Multisolvent 30
- Length:150 mm
- Packing material:Fully Porous
- Phase:PL Multisolvent 30
- Cat. No.:AGPL1515-3323
- pH Range:1 - 14
- Temperature Limit:90 °C
- Max. Pressure:2175.5 psi
- Wetted Materials:THF
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



