Order Entry
United States
ContactUsLinkComponent
Thermo Scientific Nunc DS3126-0175 Adapter/Rotor Cushion for 175 ml Bottles, PC; 4/Pk
Thermo Scientific Nunc DS3126-0175 Adapter/Rotor Cushion for 175 ml Bottles, PC; 4/Pk
Catalog # MFLX06106-78
Supplier:  Avantor
Thermo Scientific Nunc DS3126-0175 Adapter/Rotor Cushion for 175 ml Bottles, PC; 4/Pk
Catalog # MFLX06106-78
Supplier:  Avantor

About this item

Rotor cushions adapt conical-bottom tubes to rotors lacking v-bottom holders.

Cushion adds 4,3 mm to the height of the bottle.

  • Description:
    Adapter/Rotor Cushion for 175 ml Bottles, PC; 4/Pk
  • Environmentally Preferable:
  • Tube bottom shape:
    Conical
  • Tube capacity:
    175 ml
  • Cat. No.:
    MFLX06106-78
Frequently Bought Together