CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Polypropylene screens, 0.75 inch Ext. Ø for 100 ml tubes, 200-mesh, 840 µm, for BIO-DIS, Pack of 25
AG27-2200
Supplier #: 27-2200
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
AG27-2200
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
- Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
- Description:Polypropylene screens, 0.75 inch Ext. Ø for 100 ml tubes, 200-mesh, 840 µm, for BIO-DIS, Pack of 25