CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 56-mesh, 205 µm, for BIO-DIS, Pack of 25
Catalog # AG27-2007
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: 27-2007
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Catalog # AG27-2007
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
- Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Specifications
- Description:Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 56-mesh, 205 µm, for BIO-DIS, Pack of 25