Order Entry
United States
Orders LinkContactUsLinkComponent
Dissolution Sampling Cannulas

Dissolution Sampling Cannulas

Sampling cannula, 8.12 inch, for 2L vessel, for VK7000, 7010 manifold
  AG17-3307
Supplier #:  17-3307
Dissolution Sampling Cannulas
  AG17-3307
Dissolution Sampling Cannulas
Dissolution Sampling Cannulas

Sampling cannula, 8.12 inch, for 2L vessel, for VK7000, 7010 manifold

 
$64.30
   
Each
 

Available in two different lengths based on the volume of media; best used with full flow filters.

  • For manual sampling, the Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters
  • The sampling cannulas are available two in different lengths based on the volume of media: 500 or 900 ml
  • An adjustable gauge can be included with this assembly to ensure repeatable positioning for each sample

The Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters. The sampling cannulas are available in two different lengths based on the volume of media being sampled from.

 
  • Description:
    Sampling cannula, 8.12 inch, for 2L vessel, for VK7000, 7010 manifold
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.