Order Entry
United States
Orders LinkContactUsLinkComponent
Dissolution Sampling Cannulas

Dissolution Sampling Cannulas

Sampling cannula, 6.5 inch, 500 ml, for VK7000, 7010 manifold
Catalog # AG17-3305
Supplier #:  17-3305
Dissolution Sampling Cannulas
Catalog # AG17-3305
Dissolution Sampling Cannulas
Dissolution Sampling Cannulas

Sampling cannula, 6.5 inch, 500 ml, for VK7000, 7010 manifold

Price
$64.30
Log in for availability
Each
Product Details & Documents

Available in two different lengths based on the volume of media; best used with full flow filters.

  • For manual sampling, the Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters
  • The sampling cannulas are available two in different lengths based on the volume of media: 500 or 900 ml
  • An adjustable gauge can be included with this assembly to ensure repeatable positioning for each sample

The Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters. The sampling cannulas are available in two different lengths based on the volume of media being sampled from.

Specifications
  • Description:
    Sampling cannula, 6.5 inch, 500 ml, for VK7000, 7010 manifold