CrossLab Dissolution Shafts, Agilent Technologies
Basket Shaft, electropolished stainless steel, 3-clip, Conical, 21", for 705-DS; 708-DS; 709-DS; VK7000
Catalog # AG14-3624
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: 14-3624
CrossLab Dissolution Shafts, Agilent Technologies
Catalog # AG14-3624
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
Shafts of different materials and lengths, individually serialized to meet standard USP design.
- Electropolished stainless steel, PTFE-coated, gold-plated or mini basket shafts available
- Individually serialized
- Three-clip shafts to meet standard USP design
- O-ring shafts for automated systems
- Conical shafts to support air bubble dispersion
- A range of basket shaft lengths are available to accommodate various models and volume configurations
Agilent provides stainless steel, PTFE-coated, gold-plated, or mini shafts that are individually serialized. Several basket and paddle shaft lengths and options are available to meet standard USP design.
Documents
Agilent Technologies Top Supplies for 708-DS Dissolution Systems
Product Literature
Specifications
- Description:Basket Shaft, electropolished stainless steel, 3-clip, Conical, 21", for 705-DS; 708-DS; 709-DS; VK7000
- Certification:Yes
- Cat. No.:AG14-3624