Dissolution Verification Tools, Agilent Technologies
Verified Vessel centering tool, pass/fail gauge, EaseAlign centering rings, with certificate, for 705-DS; VK7000
Catalog # AG12-7310V
Jump to:
Discontinued
Product Details & Documents
A variety of different height gauges and spacers to meet all needed regulatory specifications.
- Available for the Agilent 708-DS and legacy instruments
Verification and calibration tools can help you to install your accessories properly. Agilent provides a variety of different height gauges and spacers to meet all needed regulatory specifications, such as mechanical qualification and physical parameter verification, for your dissolution test.
Specifications
- Description:Verified Vessel centering tool, pass/fail gauge, EaseAlign centering rings, with certificate, for 705-DS; VK7000
- Certification:Yes