Orders LinkContactUsLinkComponent
Dissolution Conversion Kits, Agilent Technologies
Dissolution Conversion Kits, Agilent Technologies
Conversion kit, TruCenter, with ground groove, 1000 ml, for 705-DS and VK7000/7010
Catalog # AG12-6100
Supplier #:  12-6100
Dissolution Conversion Kits, Agilent Technologies
Catalog # AG12-6100
Dissolution Conversion Kits, Agilent Technologies
Dissolution Conversion Kits, Agilent Technologies
Conversion kit, TruCenter, with ground groove, 1000 ml, for 705-DS and VK7000/7010
Price
$660.00
Log in for availability
Each
Product Details & Documents
Agilent can also support small-volume dissolution with the 708-DS dissolution apparatus.
  • Special conversion kits: available in sets of 6 and 8, also available for the Chinese Pharmacopeia (CP) 250 ml vessel
  • Conversion kits are available to upgrade your 705-DS and 7000 and 7010 models to TruCenter vessels
The Agilent 708-DS dissolution apparatus can also support small-volume dissolution. Each position will require a conversion kit to hold a 100 or 200 ml vessel in position. The conversion kits are designed to be used with either mini paddles, mini baskets, or enhancer cells.
Easy-to-install conversion kits: the 708-DS is designed to accept 100 and 200 ml vessels, the 250 ml Chinese Pharmacopoeia vessel, and standard 1 L vessels. The 2 L model can incorporate 1 and 2 L vessels as well as smaller volume vessels with conversion kits.
Specifications
  • Description:
    Conversion kit, TruCenter, with ground groove, 1000 ml, for 705-DS and VK7000/7010