Dissolution Sampling Cannulas
Bent cannula with Luer Lock and Adjustable Gauge, for sampling from 900 ml in 1 L vessels, 4.75 inch, for 708-DS, 709-DS
Catalog # AG12-3221
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: 12-3221
Jump to:
Product Details & Documents
Available in two different lengths based on the volume of media; best used with full flow filters.
- For manual sampling, the Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters
- The sampling cannulas are available two in different lengths based on the volume of media: 500 or 900 ml
- An adjustable gauge can be included with this assembly to ensure repeatable positioning for each sample
The Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters. The sampling cannulas are available in two different lengths based on the volume of media being sampled from.
Documents
Agilent Technologies Top Supplies for 708-DS Dissolution Systems
Product Literature
Specifications
- Description:Bent cannula with Luer Lock and Adjustable Gauge, for sampling from 900 ml in 1 L vessels, 4.75 inch, for 708-DS, 709-DS