Dissolution Sampling Cannulas
Bent cannula with Luer Lock and Adjustable Gauge, for sampling from 500 ml in 1 L vessels, 7.75 inch, for VK7000, 7010
AG12-3216
: 12-3216
Available in two different lengths based on the volume of media; best used with full flow filters.
- For manual sampling, the Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters
- The sampling cannulas are available two in different lengths based on the volume of media: 500 or 900 ml
- An adjustable gauge can be included with this assembly to ensure repeatable positioning for each sample
The Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters. The sampling cannulas are available in two different lengths based on the volume of media being sampled from.
- Description:Bent cannula with Luer Lock and Adjustable Gauge, for sampling from 500 ml in 1 L vessels, 7.75 inch, for VK7000, 7010