Dissolution Sampling Cannulas
Bent cannula with Luer Lock, for sampling from 900 ml in 1 L vessels, 4.75 inch, for 705-DS;708-DS;709-DS;VK7000
Catalog # AG12-3200
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: 12-3200
Jump to:
Product Details & Documents
Available in two different lengths based on the volume of media; best used with full flow filters.
- For manual sampling, the Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters
- The sampling cannulas are available two in different lengths based on the volume of media: 500 or 900 ml
- An adjustable gauge can be included with this assembly to ensure repeatable positioning for each sample
The Agilent cannula luer lock assembly is comprised of a bent cannula and a sampling syringe that are best used with the full flow filters. The sampling cannulas are available in two different lengths based on the volume of media being sampled from.
Specifications
- Description:Bent cannula with Luer Lock, for sampling from 900 ml in 1 L vessels, 4.75 inch, for 705-DS;708-DS;709-DS;VK7000