Dissolution Sinkers, Agilent Technologies
Sinker basket, Japanese Pharmacopoeia (JP), stainless steel
Catalog # AG12-3070
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: 12-3070
Dissolution Sinkers, Agilent Technologies
Catalog # AG12-3070
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
A variety of dissolution sinkers are available depending on your dosage form.
- Capsule wire, weights, and sinker baskets are used to retain a dosage form at the bottom of the vessel for testing when the dosage form is buoyant
- A variety of options are available
- The flexible capsule wire meets USP requirements and has a 0.032" diameter
- Sinker baskets are offered in a variety of mesh sizes and made of stainless steel with press-on caps, using O-rings to seal the basket
- Japanese Pharmacopoeia (JP) sinker baskets are available as an alternative
- Three-prong capsule weights fit traditional capsule sizes one through three, and are sold individually
Capsule wire, weights, and sinker baskets are used to retain a solid dosage form at the bottom of the vessel for testing when the dosage form is buoyant. Depending on your dosage form, a variety of options are available.
Documents
Agilent Technologies Top Supplies for 708-DS Dissolution Systems
Product Literature
Specifications
- Description:Sinker basket, Japanese Pharmacopoeia (JP), stainless steel