CrossLab Dissolution Baskets, Agilent Technologies
Bolus Basket, 40×40 Mesh, 381 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
Catalog # AG12-2185
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: 12-2185
CrossLab Dissolution Baskets, Agilent Technologies
Catalog # AG12-2185
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
USP 40-mesh stainless steel baskets, custom baskets, and baskets for small or large dosage forms.
- A variety of basket and mesh sizes available
- Available in different materials and coatings
- Individual Certificate of Conformity can be provided
From the standard USP 40-mesh stainless steel basket, to custom baskets with varying mesh sizes, to baskets specific to small or large dosage forms, Agilent supports your needs. Unless otherwise noted, all baskets fit the standard basket shafts.
Certifications: Individual Certificate of Conformity can be provided.
Documents
Agilent Technologies Top Supplies for 708-DS Dissolution Systems
Product Literature
Specifications
- Description:Bolus Basket, 40×40 Mesh, 381 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
- Certification:No