CrossLab Dissolution Baskets, Agilent Technologies
Basket, 325×2300 Mesh, 5 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
Catalog # AG12-2146
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Supplier #: 12-2146
CrossLab Dissolution Baskets, Agilent Technologies
Catalog # AG12-2146
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
USP 40-mesh stainless steel baskets, custom baskets, and baskets for small or large dosage forms.
- A variety of basket and mesh sizes available
- Available in different materials and coatings
- Individual Certificate of Conformity can be provided
From the standard USP 40-mesh stainless steel basket, to custom baskets with varying mesh sizes, to baskets specific to small or large dosage forms, Agilent supports your needs. Unless otherwise noted, all baskets fit the standard basket shafts.
Certifications: Individual Certificate of Conformity can be provided.
Documents
Agilent Technologies Top Supplies for 708-DS Dissolution Systems
Product Literature
Specifications
- Description:Basket, 325×2300 Mesh, 5 µm, stainless steel, for 708-DS; 709-DS; VK7000; VK7030
- Certification:No