Drawer Rack Systems for Cryogenic Freezer Storage
Product Details & Documents
Engineered for performance, organization, and durability, these stainless steel drawer rack systems provide secure and efficient sample management.
Durable stainless steel construction resists corrosion and maintains strength under ultra-cold conditions
- Versatile box compatibility; options available for 2", 3", 3.75", and mini-size box storage in plastic or cardboard formats
- Drawer-style access allows quick sample retrieval
- Compatible with a variety of cryogenic chest and upright freezer models
- Lifetime warranty on workmanship
Designed to withstand ultra-low and liquid nitrogen storage conditions, these racks combine rugged construction with convenient drawer-style access, making sample retrieval safer and faster.
Each rack is constructed from high-grade stainless steel for long-term corrosion resistance and structural integrity at cryogenic temperatures. Available in multiple configurations with different box size accommodations, these drawer racks offer flexibility for diverse lab requirements. Smooth-glide drawers and precision-welded frames ensure easy handling and consistent temperature exposure throughout the storage chamber.
Specifications
- Capacity:20 boxes
- Description:Upright stainless steel drawer rack with handles, 20 box capacity (5×4 configuration)
- Dimensions:27.54×9.12×5.49"
- For use with:2" cardboard boxes