Boron trifluoride diethyl ether complex (∼48% BF₃)
Glass bottle for solids, 1 kg
Catalog # TS17456-0010
Supplier: Thermo Scientific Chemicals
: 17456-0010
Boron trifluoride diethyl ether complex (∼48% BF₃)
Catalog # TS17456-0010
Supplier: Thermo Scientific Chemicals
CAS Number: 109-63-7
Jump to:
Product Details & Documents
Synonyms: Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
- CAS Number:109-63-7
- Formula:C₄H₁₀BF₃O
- MDL Number:MFCD00013194
- Molecular Weight:141.93 g/mol
- Merck Index:13,01340
- Boiling Pt:126 °C (760 mmHg)
- Melting Pt:–60 °C
- Density:1.12 g/cm³
- Flash Pt:58 °C
- Storage Temperature:Ambient
- EINECS:203-689-8
- UN:2604
- ADR:8,I
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:Clear colorless to straw yellow Liquid (May darken on storage)
- Refractive index:1.3429 to 1.3449 (20°C, 589 nm)
- Assay:47.5 to 49 % (BF3)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Size:1 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



