Order Entry
United States
Orders LinkContactUsLinkComponent
Phenol 99%, extra pure

Phenol 99%, extra pure

Glass bottle, 2.5 kg
Catalog # TS14934-0025
Corrosive
Toxic
Health
Environment
Supplier #:  14934-0025
Phenol 99%, extra pure
Catalog # TS14934-0025
CAS Number:  108-95-2
Phenol 99%, extra pure
Phenol 99%, extra pure

Glass bottle, 2.5 kg

CAS Number:  108-95-2
Price
$250.00
Log in for availability
Each (2,500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:182 °C (760 mmHg)
  • Melting Pt:39…42 °C
  • Density:1.07 g/cm³
  • Flash Pt:79 °C
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
  • REACH:01-2119471329-32
Specifications

Specification Test Results

  • Appearance:
    Colorless to white Crystals or fused mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    39 to 42 °C
  • Water:
    ≤0.2 %
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.