Order Entry
United States
Orders LinkContactUsLinkComponent
Buffered oxide etch 10:1 (BOE 10:1) semiconductor grade

Buffered oxide etch 10:1 (BOE 10:1) semiconductor grade

HDPE bottle, Quart
Catalog # 76541-080
Corrosive
Toxic
Health
Supplier:   Transene
Supplier #:  000BOE-QT
Buffered oxide etch 10:1 (BOE 10:1) semiconductor grade
Catalog # 76541-080
Supplier:  Transene
CAS Number:  
Buffered oxide etch 10:1 (BOE 10:1) semiconductor grade

HDPE bottle, Quart

CAS Number:  
Price
$55.81
Log in for availability
Each (1qt US)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Buffered Oxide Etchant 10:1 is a reduced-speed etchant for oxide films. 10:1 ratio of ammonium fluoride: HF.

  • Boiling Pt:103…105 °C
  • Melting Pt:–35 °C
  • Density:1.11…1.12 g/cc
  • UN:2817
  • ADR:8, 6.1,II

Documents

Specifications

Specification Test Results

  • Color:
    ≤ 10 APHA
  • Aluminum:
    ≤ 3.1 ppb
  • Antimony:
    ≤ 10 ppb
  • Arsenic:
    ≤ 2 ppb
  • Barium:
    ≤ 0.5 ppb
  • Beryllium:
    ≤ 1 ppb
  • Bismuth:
    ≤ 1 ppb
  • Boron:
    ≤ 5 ppb
  • Cadmium:
    ≤ 1 ppb
  • Calcium:
    ≤ 3 ppb
  • Chloride:
    ≤ 2 ppm
  • Chromium:
    ≤ 2 ppb
  • Cobalt:
    ≤ 2 ppb
  • Copper:
    ≤ 5 ppb
  • Gallium:
    ≤ 2 ppb
  • Germanium:
    ≤ 2 ppb
  • Gold:
    ≤ 2 ppb
  • Iron:
    ≤ 5 ppb
  • Lead:
    ≤ 2 ppb
  • Lithium:
    ≤ 1 ppb
  • Magnesium:
    ≤ 1 ppb
  • Manganese:
    ≤ 2 ppb
  • Molybdeum:
    ≤ 1 ppb
  • Nickel:
    ≤ 1 ppb
  • Nitrate:
    ≤ 3 ppm
  • Phosphate:
    ≤ 0.4 ppm
  • Potassium:
    ≤ 2 ppb
  • Silver:
    ≤ 1 ppb
  • Sodium:
    ≤ 5 ppb
  • Strontium:
    ≤ 1 ppb
  • Sulfate:
    ≤ 0.5 ppm
  • Thallium:
    ≤ 2 ppb
  • Tin:
    ≤ 2 ppb
  • Titanium:
    ≤ 5 ppb
  • Vanadium:
    ≤ 2 ppb
  • Zinc:
    ≤ 2 ppb
  • Zirconium:
    ≤ 2 ppb
  • Particle >0.5:
    ≤ 100 Particle/ml
  • Particle >1.0:
    ≤ 15 Particle/ml

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    Quart

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.