Copper etchant APS-100 semiconductor grade
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Copper Etchant APS-100 is an ammonium persulfate solution with excellent control for fine-line etching of copper films in integrated circuits.
- MDL Number:MFCD00161912
- Boiling Pt:100 °C (water)
- Density:1.08 g/cc
- UN:3139
- ADR:5.1,III
Specifications
Specification Test Results
- Appearance:White-white
- Etch Capacity (rate declines at ~70%):10 oz/gallon
- Etch Rate at 40°C:80 (etches 1oz Cu in 7.5 min) Å/sec
- Filtration:1 um
- Incompatible Materials:Al, Ni
- pH:Acidic
- Recommended Operating Temperatures:20-80°C (30-40°C most common) °C
- Shelf Life:1 Year
- Storage Conditions:Ambient
Specifications
- Pack type:HDPE bottle
- Size:Quart
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



