Potassium nitrate 99+% for analysis
Plastic bottle, 1 kg
Catalog # TS20591-0010
Supplier: Thermo Scientific Chemicals
Supplier #: AC20591-0010
Potassium nitrate 99+% for analysis
Catalog # TS20591-0010
Supplier: Thermo Scientific Chemicals
CAS Number: 7757-79-1
Jump to:
Product Details & Documents
Synonyms: Nitrate of Potash
- CAS Number:7757-79-1
- Formula:KNO₃
- MDL Number:MFCD00011409
- Molecular Weight:101.1 g/mol
- Merck Index:13,07733
- Boiling Pt:400 °C (760 mmHg)
- Melting Pt:334 °C
- Storage Temperature:Ambient
- EINECS:231-818-8
- UN:1486
- ADR:5.1,III
- REACH:01-2119488224-35
Specifications
Specification Test Results
- Appearance:White Crystals
- pH:5.5 to 8 (5 % at 20°C)
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤20 ppm
- Iodate (IO3):≤5 ppm
- Nitrite (NO2):≤10 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Sodium (Na):≤200 ppm
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Plastic bottle
- Size:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



