Lead(II) chloride 99%
Plastic bottle, 50 g
Catalog # TS19331-0500
Supplier: Thermo Scientific Chemicals
Supplier #: AC19331-0500
Lead(II) chloride 99%
Catalog # TS19331-0500
Supplier: Thermo Scientific Chemicals
CAS Number: 7758-95-4
Jump to:
Product Details & Documents
Synonyms: Lead dichloride
- CAS Number:7758-95-4
- Formula:Cl₂Pb
- MDL Number:MFCD00011157
- Molecular Weight:278.11 g/mol
- Boiling Pt:950 °C (1013 hPa)
- Melting Pt:501 °C
- Storage Temperature:Ambient
- UN:2291
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Titration Complexometric:≥98.5 %
Specifications
- Pack type:Plastic bottle
- Size:50 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



