Order Entry
United States
Orders LinkContactUsLinkComponent
Mag-Bind® Universal Pathogen Core Kit

Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 1×96 preps
Catalog # 76539-252
Supplier:   Omega Bio-Tek
Supplier #:  M4030-00
Mag-Bind® Universal Pathogen Core Kit
Catalog # 76539-252
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 1×96 preps

CAS Number:  
Price
$379.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Isolate host genomic, bacterial, fungal spore, and viral DNA and viral RNA from tissue, urine, serum, and fecal samples using magnetic beads.

  • Purified DNA is suitable for PCR, restriction digestion, and hybridization applications

The Mag-Bind® Universal Pathogen Core Kit allows rapid and reliable isolation of high-quality host genomic DNA, gram-positive and negative bacterial DNA, fungal spore DNA, viral DNA, and viral RNA from tissue, urine, serum, and fecal samples. The system incorporates a recommended enzymatic lysis which allows for fully automated extraction post-lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen Biosprint, Tecan Fluent® Automation Workstation, Tecan Freedom EVO®, and other liquid handling instruments. There are no organic extractions, thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel. The system combines the Mag-Bind® technology with RBB Buffer to eliminate PCR inhibiting compounds within the samples. Purified DNA is suitable for PCR, restriction digestion, and hybridization applications.

Specifications

Specifications

  • Description:
    Universal pathogen kit
  • Size:
    1×96 preps
  • Cat. No.:
    76539-252
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Sample type:
    Bacteria

Specifications