Acetone, Technical Grade
Plastic drum, 5 L
Catalog # TS44415-0050
Supplier: Thermo Scientific Chemicals
Supplier #: 44415-0050
Acetone, Technical Grade
Catalog # TS44415-0050
Supplier: Thermo Scientific Chemicals
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Boiling Pt:56 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- UN:1090
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Refractive index:1.3580 to 1.3600 (20°C, 589 nm)
- Density:0.790 to 0.792 kg/l
- GC:≥98.5 %
Specifications
- Pack type:Plastic drum
- Size:5 L
- Cat. No.:TS44415-0050
- Environmental Certification:Chem21 Solvent selection guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



