Cycloheximide 95%
Glass bottle for solids, 1 g
Catalog # TS35742-0010
Supplier: Thermo Scientific Chemicals
Supplier #: 35742-0010
Jump to:
Product Details & Documents
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Number:66-81-9
- Formula:C₁₅H₂₃NO₄
- MDL Number:MFCD00082346
- Molecular Weight:281.35
- Melting Pt:107…114 °C
- Storage Temperature:Ambient
- EINECS:200-636-0
- UN:2811
- ADR:6.1,I
Specifications
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Melting point:108 to 118 °C
- HPLC:≥94 %
- Specific optical rotation:-25° to -32° (20 °C, 589 nm) (c=1, CHCl3)
Specifications
- Pack type:Glass bottle for solids
- Size:1 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



