Order Entry
United States
Orders LinkContactUsLinkComponent
D-(+)-Mannose 99+%

D-(+)-Mannose 99+%

Plastic bottle, 100 g
Catalog # TS15060-1000
Supplier #:  15060-1000
D-(+)-Mannose 99+%
Catalog # TS15060-1000
CAS Number:  3458-28-4
D-(+)-Mannose 99+%
D-(+)-Mannose 99+%

Plastic bottle, 100 g

CAS Number:  3458-28-4
Price
$184.00
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Melting Pt:133 °C
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.45 %
  • HPLC:
    ≥99.0 %
  • Appearance of solution:
    (10 % in water) Clear
  • Bacterial endotoxins:
    (LAL-test) 0.5 E.U. in 1 ml of a 0.1% soln.
  • Specific optical rotation:
    +13.5° to +14.9° (20°C, 589 nm) (c=10, water) (after 24 hours, on dry substance)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.