BUFFER ACIDIC LYSIS 1 100ML ROOM TEMP
Catalog # MSPP-910-0010
Supplier: CHEMOMETEC INC MS
BUFFER ACIDIC LYSIS 1 100ML ROOM TEMP
Catalog # MSPP-910-0010
Supplier: CHEMOMETEC INC MS
Supplier Number: 910-0010
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
CASSETTE VIA2 PK100
Supplier: CHEMOMETEC INC MS
Sol-Care® Safety Needles
Supplier: SOL-MILLENNIUM MEDICAL
Human Recombinant DLL3 (from HEK293), FITC Labeled Protein
Supplier: ACROBIOSYSTEMS