ContactUsLinkComponent
Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for High-Performance Precision Tubing, SS
Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for High-Performance Precision Tubing, SS
Catalog # MFLX77200-69
undefined
Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for High-Performance Precision Tubing, SS
Catalog # MFLX77200-69
Supplier Number:  77200-69
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • Currently Viewing
Specifications
About this item
Get the most our of your Masterflex® L/S® Easy-Load® II Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® pump head
  • Suitable for use with Easy-Load® II pump head 77200-62
Replace your pump head rotor assembly to extend the life of your pump head.
  • Pump series:
    L/S
  • Description:
    Replacement rotor assembly for L/S Easy-Load II pump heads 77200-62,
  • Environmentally Preferable:
  • Warranty:
    2 years
  • Cat. No.:
    MFLX77200-69
  • Rotor Material:
    Stainless steel
Frequently Bought Together