ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Y-Connector, Hose Barb Union, ¹/₈" Int.Ø; 100 Pack
Masterflex® Fitting, Polypropylene, Y-Connector, Hose Barb Union, ¹/₈" Int.Ø; 100 Pack
Catalog # MFLX40727-43
Supplier:  VWR International
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Y-Connector, Hose Barb Union, ¹/₈" Int.Ø; 100 Pack
Catalog # MFLX40727-43
Supplier:  VWR International
CE certificate
About this item
No tools needed for assembly.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: 1/8" ID; 100/PK
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
  • Barb size/tubing int.Ø (port 1):
    ⅛"
  • Barb size/tubing int.Ø (port 2):
    ⅛"
  • Connection Type:
    Barbed Union
  • Flow pattern:
    Y
  • Description:
    Fitting, polypropylene, Y-connector, hose barb union, ¹/₈" Int.Ø; 100 pack
  • Environmentally Preferable:
  • Model:
    40727-43
  • Sterilization:
    Gamma Irradiation, Autoclave, and EtO
  • Warranty:
    3 Months
  • Cat. No.:
    MFLX40727-43
  • Fitting Material:
    PP
  • Barb Size / Tubing I.D. (Port 3):
    ⅛"
Frequently Bought Together