ContactUsLinkComponent
Compression male pipe adapter elbow, 3/8" NPT(M)×3/8" OD, brass
Compression male pipe adapter elbow, 3/8" NPT(M)×3/8" OD, brass
Catalog # MFLX31412-86
undefined
Compression male pipe adapter elbow, 3/8" NPT(M)×3/8" OD, brass
Catalog # MFLX31412-86
Supplier Number:  IBD6ME6
  • Warranty:
  • LTD life time
  • Currently Viewing
Specifications
About this item
Maintain a permanent connection with these adapters.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    Elbow
  • Thread size NPT(M) (port 2):
    ⅜"
  • Description:
    Compression male pipe adapter elbow, 3/8" NPT(M)×3/8" OD, brass
  • Environmentally Preferable:
  • Warranty:
    LTD life time
  • Cat. No.:
    MFLX31412-86
  • Fitting Material:
    Brass
Frequently Bought Together