FMI P56C Series Q pump head to NEMA type 56 C-face motor adapter kit
Catalog # MFLX07117-85
Supplier: Avantor
About this item
Configurable to your requirements.
- Easily adapt and mount your Series Q pump head
Operate the Series Q pump head (sold on the opposite page) with Masterflex L/S™ drives (sold on pages 787-806) and NEMA Type 56C-face motors (sold on pages 897-899) using these adapter kits.
- Description:Series Q pump head to NEMA type 56 C-face motor adapter kit
- Environmentally Preferable:
- Warranty:1 year
- Cat. No.:MFLX07117-85
Frequently Bought Together